2-Fluoro-6-methoxy-4-methylphenylboronic acid pinacol ester ≥95%

Supplier: AOBChem USA

Warning

33508-5G 33508-1G 33508-25G 33508-500MG
77632-808EA 885 USD
77632-808 77638-168 77655-148 77646-146
2-Fluoro-6-methoxy-4-methylphenylboronic acid pinacol ester ≥95%
2-Fluoro-6-methoxy-4-methylphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₄H₂₀BFO₃
Storage Temperature: Ambient
MDL Number: MFCD30834072
CAS Number: 2121513-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR