Methyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥97%

Supplier: AOBChem USA

Warning

33497-5G 33497-25G 33497-1G 33497-500MG
77632-806EA 1198 USD
77632-806 77655-146 77642-150 77646-144
Methyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate ≥97%
Methyl 3-bromo-2-fluoro-5-(trifluoromethyl)benzoate

Precaución: For Research Use Only

Formula: C₉H₅BrF₄O₂
MW: 301.032 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30834068
CAS Number: 537658-03-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR