4,5-Difluoro-2-(propan-2-yloxy)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

33489-5G 33489-25G 33489-1G 33489-500MG
77632-802EA 878 USD
77632-802 77658-804 77642-146 77650-178
4,5-Difluoro-2-(propan-2-yloxy)benzoic acid ≥97%
4,5-Difluoro-2-(propan-2-yloxy)benzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₀F₂O₃
Storage Temperature: Ambient
MDL Number: MFCD28627593
CAS Number: 1782612-50-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR