2-(Benzyloxy)-4,5-difluorobenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

33483-5G 33483-25G 33483-1G 33483-500MG
77634-816EA 536 USD
77634-816 77655-142 77642-142 77646-142
2-(Benzyloxy)-4,5-difluorobenzaldehyde ≥97%
2-(Benzyloxy)-4,5-difluorobenzaldehyde

Precaución: For Research Use Only

Formula: 248.225 g/mol
MW: C₁₄H₁₀F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD26960223
CAS Number: 1823269-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR