1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

334563-5G 334563-1G 334563-25G 334563-500MG
77634-814EA 348 USD
77634-814 77638-164 77655-138 77646-140
1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene ≥97%
1-Bromo-2-(2,2-diethoxyethoxy)-4-fluorobenzene

Precaución: For Research Use Only

Formula: C₁₂H₁₆BrFO₃
MW: 307.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13191631
CAS Number: 253429-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR