[5-Chloro-2-(propan-2-yloxy)pyridin-4-yl]methanol ≥97%

Supplier: AOBChem USA

Warning

33441-5G 33441-1G 33441-25G 33441-500MG
77632-798EA 941 USD
77632-798 77638-162 77658-802 77646-136
[5-Chloro-2-(propan-2-yloxy)pyridin-4-yl]methanol ≥97%
[5-Chloro-2-(propan-2-yloxy)pyridin-4-yl]methanol

Precaución: For Research Use Only

Formula: 201.65 g/mol
MW: C₉H₁₂ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD30298595
CAS Number: 1602043-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR