Methyl 5-chloro-2-(propan-2-yloxy)pyridine-4-carboxylate ≥97%

Supplier: AOBChem USA

Warning

33439-5G 33439-1G 33439-500MG
77658-800EA 2011 USD
77658-800 77642-138 77650-172
Methyl 5-chloro-2-(propan-2-yloxy)pyridine-4-carboxylate ≥97%
Methyl 5-chloro-2-(propan-2-yloxy)pyridine-4-carboxylate

Precaución: For Research Use Only

Formula: 229.66 g/mol
MW: C₁₀H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD30834064
CAS Number: 2121515-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR