1-Bromo-2,3,4-trichlorobenzene ≥95%

Supplier: AOBChem USA

Warning

33435-5G 33435-500MG 33435-25G 33435-1G
77632-796EA 683 USD
77632-796 77652-682 77658-798 77646-134
1-Bromo-2,3,4-trichlorobenzene ≥95%
1-Bromo-2,3,4-trichlorobenzene

Precaución: For Research Use Only

Formula: 260.343 g/mol
MW: C₆H₂BrCl₃
Storage Temperature: Ambient
MDL Number: MFCD01026473
CAS Number: 81067-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR