3-Bromo-5-chloro-4-hydroxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

333927-1G 333927-500MG 333927-5G
77638-156EA 539 USD
77638-156 77652-680 77658-792
3-Bromo-5-chloro-4-hydroxybenzonitrile ≥97%
3-Bromo-5-chloro-4-hydroxybenzonitrile

Precaución: For Research Use Only

Formula: 232.46 g/mol
MW: C₇H₃BrClNO
Storage Temperature: Ambient
MDL Number: MFCD22054958
CAS Number: 1689-86-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR