3-Chloro-2-methoxypyridin-4-ol ≥97%

Supplier: AOBChem USA

Warning

33376-5G 33376-1G 33376-25G 33376-500MG
77632-790EA 731 USD
77632-790 77638-154 77658-790 77650-164
3-Chloro-2-methoxypyridin-4-ol ≥97%
3-Chloro-2-methoxypyridin-4-ol

Precaución: For Research Use Only

Formula: 159.57 g/mol
MW: C₆H₆ClNO₂
Storage Temperature: Ambient
MDL Number: MFCD16610466
CAS Number: 1227600-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR