(2-Chloro-4-methylpyridin-3-yl)methanol ≥95%

Supplier: AOBChem USA

Warning

33352-5G 33352-1G 33352-500MG 33352-25G
77632-788EA 1330 USD
77632-788 77638-152 77652-678 77658-788
(2-Chloro-4-methylpyridin-3-yl)methanol ≥95%
(2-Chloro-4-methylpyridin-3-yl)methanol

Precaución: For Research Use Only

Formula: 157.598 g/mol
MW: C₇H₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD16610706
CAS Number: 447402-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR