2,4-Di-tert-butyl-1-methoxybenzene ≥97%

Supplier: AOBChem USA

Warning

332892-5G 332892-500MG 332892-25G 332892-1G
77634-808EA 566 USD
77634-808 77652-676 77655-132 77642-136
2,4-Di-tert-butyl-1-methoxybenzene ≥97%
2,4-Di-tert-butyl-1-methoxybenzene

Precaución: For Research Use Only

Formula: 220.35 g/mol
MW: C₁₅H₂₄O
Storage Temperature: Ambient
MDL Number: MFCD24038836
CAS Number: 17177-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR