[2-Bromo-6-(methylsulfanyl)phenyl]methanol ≥97%

Supplier: AOBChem USA

Warning

33288-5G 33288-1G 33288-25G 33288-500MG
77632-786EA 933 USD
77632-786 77638-148 77655-130 77650-160
[2-Bromo-6-(methylsulfanyl)phenyl]methanol ≥97%
[2-Bromo-6-(methylsulphanyl)phenyl]methanol

Precaución: For Research Use Only

Formula: 233.125 g/mol
MW: C₈H₉BrOS
Storage Temperature: Ambient
MDL Number: MFCD30737499
CAS Number: 1898133-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR