2-Iododiphenylmethane ≥97%

Supplier: AOBChem USA

Warning

332700-5G 332700-25G 332700-1G 332700-500MG
77632-782EA 1061 USD
77632-782 77655-128 77642-132 77650-156
2-Iododiphenylmethane ≥97%
2-Iododiphenylmethane

Precaución: For Research Use Only

Formula: C₁₃H₁₁I
Storage Temperature: Ambient
MDL Number: MFCD00270110
CAS Number: 35444-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR