4-Fluoro-3-isopropoxybenzoic acid methyl ester ≥97%

Supplier: AOBChem USA

Warning

33246-5G 33246-25G 33246-1G 33246-500MG
77634-806EA 469 USD
77634-806 77655-124 77642-128 77650-152
4-Fluoro-3-isopropoxybenzoic acid methyl ester ≥97%
4-Fluoro-3-isopropoxybenzoic acid methyl ester

Precaución: For Research Use Only

Formula: 212.218 g/mol
MW: C₁₁H₁₃FO₃
Storage Temperature: Ambient
MDL Number: MFCD23115006
CAS Number: 869990-48-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR