3-(4-Bromo-1H-pyrazol-1-yl)propanenitrile ≥97%

Supplier: AOBChem USA

Warning

33217-5G 33217-25G 33217-1G 33217-500MG
77632-778EA 670 USD
77632-778 77658-782 77642-126 77646-126
3-(4-Bromo-1H-pyrazol-1-yl)propanenitrile ≥97%
3-(4-Bromo-1H-pyrazol-1-yl)propanenitrile

Precaución: For Research Use Only

Formula: C₆H₆BrN₃
Storage Temperature: Ambient
MDL Number: MFCD08701108
CAS Number: 1007518-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR