1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone ≥95%

Supplier: AOBChem USA

Warning

332018-5G 332018-1G 332018-500MG
77628-652EA 1484.23 USD
77628-652 77642-122 77650-146
1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone ≥95%
1-[6-(4-Chlorophenyl)pyridin-3-yl]ethanone

Precaución: For Research Use Only

Formula: C₁₃H₁₀ClNO
MW: 231.68 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12404912
CAS Number: 1216474-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR