3,4-Difluoro-5-ethoxyphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33184-5G 33184-1G 33184-25G 33184-500MG
77632-776EA 656 USD
77632-776 77638-144 77658-780 77650-144
3,4-Difluoro-5-ethoxyphenylboronic acid pinacol ester ≥97%
3,4-Difluoro-5-ethoxyphenylboronic acid pinacol ester

Precaución: For Research Use Only

Formula: 284.107 g/mol
MW: C₁₄H₁₉BF₂O₃
Storage Temperature: Ambient
MDL Number: MFCD30723178
CAS Number: 2121512-90-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR