2-Isopropyl-4-methoxyphenyl boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

33176-5G 33176-1G 33176-500MG
77655-118EA 2611 USD
77655-118 77642-120 77650-142
2-Isopropyl-4-methoxyphenyl boronic acid pinacol ester ≥97%
2-Isopropyl-4-methoxyphenyl boronic acid pinacol ester

Precaución: For Research Use Only

Formula: C₁₆H₂₅BO₃
Storage Temperature: Ambient
MDL Number: MFCD29105172
CAS Number: 2121512-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR