3,4-Dichloro-2-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

33153-5G 33153-1G 33153-25G 33153-500MG
77632-774EA 933 USD
77632-774 77638-142 77658-778 77646-122
3,4-Dichloro-2-methoxyphenol ≥97%
3,4-Dichloro-2-methoxyphenol

Precaución: For Research Use Only

Formula: C₇H₆Cl₂O₂
Storage Temperature: Ambient
MDL Number: MFCD24713701
CAS Number: 77102-94-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR