[2,2'-Bithiophen]-5-yl(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

331447-5G 331447-1G 331447-500MG 331447-25G
77628-650EA 1484.23 USD
77628-650 77638-140 77652-674 77658-776
[2,2'-Bithiophen]-5-yl(cyclopropyl)methanol ≥95%
[2,2'-Bithiophen]-5-yl(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: C₁₂H₁₂OS₂
Storage Temperature: Ambient
MDL Number: MFCD35181268
CAS Number: 3002537-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR