2,2-Dichloro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

331430-5G 331430-1G 331430-500MG 331430-25G
77628-646EA 1484.23 USD
77628-646 77638-136 77652-672 77655-116
2,2-Dichloro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone ≥95%
2,2-Dichloro-1-(2-fluoro-3-(trifluoromethyl)phenyl)ethanone

Precaución: For Research Use Only

Formula: 275.027 g/mol
MW: C₉H₄Cl₂F₄O
Storage Temperature: Ambient
MDL Number: MFCD35181253
CAS Number: 3002477-15-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR