2,5-Dimethoxy-4-methylbenzoic acid ≥95%

Supplier: AOBChem USA

Warning

33135-5G 33135-1G 33135-25G 33135-500MG
77632-772EA 701 USD
77632-772 77638-134 77655-114 77650-138
2,5-Dimethoxy-4-methylbenzoic acid ≥95%
2,5-Dimethoxy-4-methylbenzoic acid

Precaución: For Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD13704861
CAS Number: 32176-94-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR