1-(4-Isobutoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%

Supplier: AOBChem USA

Warning

331159-5G 331159-1G 331159-25G 331159-500MG
77628-644EA 1484.23 USD
77628-644 77638-130 77658-774 77646-120
1-(4-Isobutoxyphenyl)-2,2-dimethylpropan-1-ol ≥95%
1-(4-Isobutoxyphenyl)-2,2-dimethylpropan-1-ol

Precaución: For Research Use Only

Formula: C₁₅H₂₄O₂
Storage Temperature: Ambient
MDL Number: MFCD35181021
CAS Number: 3002480-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR