Cyclohexyl(4-propoxyphenyl)methanone ≥97%

Supplier: AOBChem USA

Warning

331140-5G 331140-25G 331140-1G 331140-500MG
77628-642EA 1484.23 USD
77628-642 77635-652 77638-128 77646-116
Cyclohexyl(4-propoxyphenyl)methanone ≥97%
Cyclohexyl(4-propoxyphenyl)methanone

Precaución: For Research Use Only

Formula: C₁₆H₂₂O₂
Storage Temperature: Ambient
MDL Number: MFCD16324255
CAS Number: 1225732-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR