2-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%

Supplier: AOBChem USA

Warning

331078-5G 331078-1G 331078-500MG
77655-110EA 1898 USD
77655-110 77642-116 77650-134
2-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol ≥95%
2-(3-Methoxy-5-(trifluoromethyl)phenyl)propan-2-ol

Precaución: For Research Use Only

Formula: 234.215 g/mol
MW: C₁₁H₁₃F₃O₂
Storage Temperature: Ambient
MDL Number: MFCD35180957
CAS Number: 1783618-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR