1-Iodo-5-isopropyl-4-methoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

330905-5G 330905-25G 330905-1G 330905-500MG
77628-640EA 1484.23 USD
77628-640 77658-770 77642-114 77650-132
1-Iodo-5-isopropyl-4-methoxy-2-methylbenzene ≥95%
1-Iodo-5-isopropyl-4-methoxy-2-methylbenzene

Precaución: For Research Use Only

Formula: 290.141 g/mol
MW: C₁₁H₁₅IO
Storage Temperature: Ambient
MDL Number: MFCD30329563
CAS Number: 1195245-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR