(5-Isopropyl-4-methoxy-2-methylphenyl)(methyl)sulfane ≥95%
Supplier: AOBChem USA
|
Warning
|
330903-5G
330903-1G
330903-25G
330903-500MG
77628-638EA
1484.23
USD
77628-638
77638-126
77655-108
77650-130
(5-Isopropyl-4-methoxy-2-methylphenyl)(methyl)sulfane ≥95%
(5-Isopropyl-4-methoxy-2-methylphenyl)(methyl)sulphane
Precaución: For Research Use Only
Formula:
210.336 g/mol MW: C₁₂H₁₈OS Storage Temperature: Ambient |
MDL Number:
MFCD35180799 CAS Number: 3002490-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...