3-Bromo-5-chloro-2-ethoxypyridine ≥97%

Supplier: AOBChem USA

Warning

33083-5G 33083-25G 33083-1G 33083-500MG
77632-768EA 841 USD
77632-768 77655-102 77642-110 77650-126
3-Bromo-5-chloro-2-ethoxypyridine ≥97%
3-Bromo-5-chloro-2-ethoxypyridine

Precaución: For Research Use Only

Formula: 236.494 g/mol
MW: C₇H₇BrClNO
Storage Temperature: Ambient
MDL Number: MFCD18257585
CAS Number: 943153-41-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR