5-Bromo-2-methyl-2H-1,2,3-triazole-4-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

330591-1G 330591-500MG 330591-5G
77638-124EA 760 USD
77638-124 77652-670 77658-768
5-Bromo-2-methyl-2H-1,2,3-triazole-4-carboxaldehyde ≥97%
5-Bromo-2-methyl-2H-1,2,3-triazole-4-carboxaldehyde

Precaución: For Research Use Only

Formula: C₄H₄BrN₃O
Storage Temperature: Ambient
MDL Number: MFCD31556386
CAS Number: 1306610-18-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR