2-Chloro-α-phenyl-5-(trifluoromethyl)benzenemethanol ≥95%

Supplier: AOBChem USA

Warning

330580-5G 330580-1G 330580-25G 330580-500MG
77628-634EA 1484.23 USD
77628-634 77638-122 77658-766 77646-114
2-Chloro-α-phenyl-5-(trifluoromethyl)benzenemethanol ≥95%
2-Chloro-α-phenyl-5-(trifluoromethyl)benzenemethanol

Precaución: For Research Use Only

Formula: 286.68 g/mol
MW: C₁₄H₁₀ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD35180626
CAS Number: 118286-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR