2-Chloro-α-ethenyl-3,6-difluorobenzenemethanol ≥95%

Supplier: AOBChem USA

Warning

330458-5G 330458-1G 330458-500MG
77658-764EA 2001 USD
77658-764 77642-108 77646-112
2-Chloro-α-ethenyl-3,6-difluorobenzenemethanol ≥95%
2-Chloro-α-ethenyl-3,6-difluorobenzenemethanol

Precaución: For Research Use Only

Formula: C₉H₇ClF₂O
Storage Temperature: Ambient
MDL Number: MFCD30628670
CAS Number: 1175300-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR