5-(Methoxymethyl)pyridine-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

33045-5G 33045-1G 33045-500MG
77655-100EA 1836 USD
77655-100 77642-106 77646-110
5-(Methoxymethyl)pyridine-3-carboxylic acid ≥97%
5-(Methoxymethyl)pyridine-3-carboxylic acid

Precaución: For Research Use Only

Formula: 167.162 g/mol
MW: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD18257073
CAS Number: 1000298-38-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR