3-Bromo-2-ethoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

32962-5G 32962-1G 32962-500MG 32962-25G
77632-766EA 642 USD
77632-766 77638-120 77652-668 77658-762
3-Bromo-2-ethoxybenzaldehyde ≥97%
3-Bromo-2-ethoxybenzaldehyde

Precaución: For Research Use Only

Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD14695221
CAS Number: 1009091-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR