1-(4-Bromobenzyl)-4-methylpiperidine ≥97%

Supplier: AOBChem USA

Warning

32890-5G 32890-25G 32890-1G 32890-500MG
77634-796EA 260 USD
77634-796 77658-756 77642-102 77650-120
1-(4-Bromobenzyl)-4-methylpiperidine ≥97%
1-(4-Bromobenzyl)-4-methylpiperidine

Precaución: For Research Use Only

Formula: 268.193 g/mol
MW: C₁₃H₁₈BrN
Storage Temperature: Ambient
MDL Number: MFCD06081265
CAS Number: 891403-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR