1-(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
Supplier: AOBChem USA
|
Warning
|
328825-5G
328825-25G
328825-1G
328825-500MG
77628-630EA
1484.23
USD
77628-630
77655-096
77642-100
77650-118
1-(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)-2,2,2-trifluoroethanone ≥95%
1-(4-(Cyclopropylmethoxy)-2,6-difluorophenyl)-2,2,2-trifluoroethanone
Precaución: For Research Use Only
Formula:
280.191 g/mol MW: C₁₂H₉F₅O₂ Storage Temperature: Ambient |
MDL Number:
MFCD35180187 CAS Number: 3002537-39-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...