Methyl 2-bromo-6-fluoro-3-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

32861-5G 32861-25G 32861-1G 32861-500MG
77634-794EA 370 USD
77634-794 77655-094 77642-098 77650-116
Methyl 2-bromo-6-fluoro-3-methylbenzoate ≥97%
Methyl 2-bromo-6-fluoro-3-methylbenzoate

Precaución: For Research Use Only

Formula: C₉H₈BrFO₂
Storage Temperature: Ambient
MDL Number: MFCD22987925
CAS Number: 1359857-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR