Phenol, 4-iodo-, 1-methanesulfonate ≥97%

Supplier: AOBChem USA

Warning

328551-5G 328551-25G 328551-1G 328551-500MG
77632-762EA 1381 USD
77632-762 77655-092 77642-096 77650-114
Phenol, 4-iodo-, 1-methanesulfonate ≥97%
Phenol, 4-iodo-, 1-methanesulphonate

Precaución: For Research Use Only

Formula: 298.1 g/mol
MW: C₇H₇IO₃S
Storage Temperature: Ambient
MDL Number: MFCD00805375
CAS Number: 291314-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR