Phenol, 3-bromo-, 1-methanesulfonate ≥97%

Supplier: AOBChem USA

Warning

328529-5G 328529-25G 328529-1G 328529-500MG
77632-760EA 986 USD
77632-760 77658-754 77642-094 77650-112
Phenol, 3-bromo-, 1-methanesulfonate ≥97%
Phenol, 3-bromo-, 1-methanesulphonate

Precaución: For Research Use Only

Formula: C₇H₇BrO₃S
Storage Temperature: Ambient
MDL Number: MFCD16036339
CAS Number: 219946-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR