5-Ethoxy-2-fluoro-4-iodopyridine ≥97%

Supplier: AOBChem USA

Warning

32786-5G 32786-1G 32786-500MG
77628-626EA 1484.23 USD
77628-626 77638-114 77650-110
5-Ethoxy-2-fluoro-4-iodopyridine ≥97%
5-Ethoxy-2-fluoro-4-iodopyridine

Precaución: For Research Use Only

Formula: 267.039 g/mol
MW: C₇H₇FINO
Storage Temperature: Ambient
MDL Number: MFCD28795550
CAS Number: 1805067-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR