4-tert-Butylbenzophenone ≥97%

Supplier: AOBChem USA

Warning

327697-5G 327697-25G 327697-1G
77634-792EA 245 USD
77634-792 77655-090 77642-090
4-tert-Butylbenzophenone ≥97%
4-tert-Butylbenzophenone

Precaución: For Research Use Only

Formula: C₁₇H₁₈O
MW: 238.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00017255
CAS Number: 22679-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR