3-Chloro-2-methoxy-4-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

32767-5G 32767-1G 32767-25G 32767-500MG
77634-790EA 492 USD
77634-790 77638-112 77655-088 77650-108
3-Chloro-2-methoxy-4-methylbenzoic acid ≥97%
3-Chloro-2-methoxy-4-methylbenzoic acid

Precaución: For Research Use Only

Formula: 200.619 g/mol
MW: C₉H₉ClO₃
Storage Temperature: Ambient
MDL Number: MFCD26671416
CAS Number: 1537639-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR