1-(Phenylmethyl)-2-(trimethylsilyl)benzene 95

Supplier: AOBChem USA

Warning

327614-5G 327614-1G 327614-500MG 327614-25G
77628-624EA 1484.23 USD
77628-624 77638-110 77652-666 77655-086
1-(Phenylmethyl)-2-(trimethylsilyl)benzene 95
1-(Phenylmethyl)-2-(trimethylsilyl)benzene

Precaución: For Research Use Only

Formula: C₁₆H₂₀Si
MW: 240.42 g/mol
Storage Temperature: Ambient
MDL Number: MFCD35179480
CAS Number: 33356-47-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR