2-Chloro-N-[2-(phenylmethyl)phenyl]acetamide ≥95%

Supplier: AOBChem USA

Warning

327594-5G 327594-25G 327594-1G 327594-500MG
77632-758EA 1113 USD
77632-758 77655-084 77642-088 77650-106
2-Chloro-N-[2-(phenylmethyl)phenyl]acetamide ≥95%
2-Chloro-N-[2-(phenylmethyl)phenyl]acetamide

Precaución: For Research Use Only

Formula: 259.73 g/mol
MW: C₁₅H₁₄ClNO
Storage Temperature: Ambient
CAS Number: 21535-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR