Phenyl(2,4,6-trichlorophenyl)methanone ≥95%

Supplier: AOBChem USA

Warning

327354-5G 327354-25G 327354-1G 327354-500MG
77628-620EA 1484.23 USD
77628-620 77635-646 77638-108 77646-100
Phenyl(2,4,6-trichlorophenyl)methanone ≥95%
Phenyl(2,4,6-trichlorophenyl)methanone

Precaución: For Research Use Only

Formula: C₁₃H₇Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD00156817
CAS Number: 25187-07-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR