1-(2,4,6-Trichlorophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

327353-1G 327353-5G 327353-500MG
77638-106EA 655 USD
77638-106 77658-752 77650-104
1-(2,4,6-Trichlorophenyl)ethanone ≥97%
1-(2,4,6-Trichlorophenyl)ethanone

Precaución: For Research Use Only

Formula: C₈H₅Cl₃O
Storage Temperature: Ambient
MDL Number: MFCD19707600
CAS Number: 52120-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR