(3-Chloro-2-fluoro-5-methylphenyl)(phenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327327-5G 327327-25G 327327-1G 327327-500MG
77632-756EA 1441 USD
77632-756 77655-082 77642-084 77646-098
(3-Chloro-2-fluoro-5-methylphenyl)(phenyl)methanol ≥95%
(3-Chloro-2-fluoro-5-methylphenyl)(phenyl)methanol

Precaución: For Research Use Only

Formula: 250.696 g/mol
MW: C₁₄H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD35179394
CAS Number: 3002437-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR