1-(3-Chloro-2-fluoro-5-methylphenyl)cyclopentanol ≥95%
Supplier: AOBChem USA
|
Warning
|
327325-5G
327325-1G
327325-25G
327325-500MG
77628-616EA
1484.23
USD
77628-616
77638-102
77655-080
77646-096
1-(3-Chloro-2-fluoro-5-methylphenyl)cyclopentanol ≥95%
1-(3-Chloro-2-fluoro-5-methylphenyl)cyclopentanol
Precaución: For Research Use Only
Formula:
228.69 g/mol MW: C₁₂H₁₄ClFO Storage Temperature: Ambient |
MDL Number:
MFCD35179392 CAS Number: 3002534-26-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...