(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanol ≥95%

Supplier: AOBChem USA

Warning

327323-5G 327323-25G 327323-1G 327323-500MG
77632-754EA 1441 USD
77632-754 77655-078 77642-082 77646-094
(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanol ≥95%
(3-Chloro-2-fluoro-5-methylphenyl)(cyclopropyl)methanol

Precaución: For Research Use Only

Formula: 214.664 g/mol
MW: C₁₁H₁₂ClFO
Storage Temperature: Ambient
MDL Number: MFCD35179390
CAS Number: 3002422-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR