1-(3-Chloro-2-fluoro-5-methylphenyl)propan-1-one ≥95%

Supplier: AOBChem USA

Warning

327321-5G 327321-1G 327321-25G 327321-500MG
77628-614EA 1484.23 USD
77628-614 77638-100 77658-750 77646-092
1-(3-Chloro-2-fluoro-5-methylphenyl)propan-1-one ≥95%
1-(3-Chloro-2-fluoro-5-methylphenyl)propan-1-one

Precaución: For Research Use Only

Formula: 200.637 g/mol
MW: C₁₀H₁₀ClFO
Storage Temperature: Ambient
MDL Number: MFCD35179388
CAS Number: 3002534-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR